Assay Details
Gene Symbol: Krtap6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010672.3 | NP_034802.1 | 1 | 451 | 95 | |
GenBank mRNA | BC107313.1 | - | 1 | 451 | 40053340 | 95 |
BC107314.1 | - | 1 | 451 | 40053341 | 95 | |
BC114404.1 | - | 1 | 449 | 95 | ||
BC114532.1 | - | 1 | 449 | 95 | ||
BC127045.1 | - | 1 | 451 | 95 | ||
BC127046.1 | - | 1 | 451 | 95 | ||
CJ048388.1 | - | 1 | 491 | 95 | ||
D86420.1 | - | 1 | 451 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119764 | AK003924.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095054 | NM_010672.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119764 | AK003924.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095054 | NM_010672.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949301 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949314 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949307 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949325 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949305 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949310 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949308 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949303 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949320 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30949312 | - | - |
Whole Mouse Genome G4122A | A_52_P479051 | D86421.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P479051 | AK137757.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12748225 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12748235 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12748238 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12748229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30949341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12658034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12748233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949354 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12565219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12815232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30949337 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10436761 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451859_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10436760 | - | - |
Mouse Genome 430 2.0 Array | 1451859_at | - | - |
Mouse Genome 430 2.0 Array | 1425872_at | - | - |
Mouse Genome 430 2.0 Array | 1420751_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420751_at | - | - |
Mouse Genome 430 2.0 Array | 1425872_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451859_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16700_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16700_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600007586.mm.1 | NM_010672.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600007586.mm.1 | NM_010672.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600007586.mm.1 | TC1600000968.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420751_PM_at | - | - |
Mouse Expression Set 430 | 1451859_at | - | - |
Mouse Expression Set 430 | 1420751_at | - | - |
Mouse Expression Set 430 | 1451859_PM_at | - | - |
Murine Genome U74v2 Set | 101374_at | - | - |
Murine Genome U74v2 Set | 101374_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420751_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451859_at | - | - |
Mouse Genome 430A 2.0 Array | 1451859_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420751_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420751_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451859_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451859_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420751_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2100193 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103440593 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2100193 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106650497 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102350184 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002100193 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694230 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1236523 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1236905 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2694230 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2438697 | - | - |
mouse-6 v1.1 (Illumina) | scl1155.1.1_180-S | - | - |
mouse-6 v1.1 (Illumina) | scl400.1.1_264-S | - | - |
mouse-6 v1.1 (Illumina) | scl48901.1.20_213-S | - | - |
mouse-6 v1.1 (Illumina) | scl399.1.1_127-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2100193 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103440593 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106650497 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102350184 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl48901.1.20_213-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102350184 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106650497 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103440593 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100193 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2694230 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2438697 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694230 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236523 | - | - |
