Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017457.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017457.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017457.mm.1 |
TC0400002093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017458.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017458.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017458.mm.1 |
TC0400002093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017459.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017459.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017459.mm.1 |
TC0400002093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017460.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017460.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017460.mm.1 |
TC0400002093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017464.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017464.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017464.mm.1 |
TC0400002093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017468.mm.1 |
NM_025338.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017468.mm.1 |
NM_025338.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017468.mm.1 |
TC0400002093.mm.1 |
MTA TC |