Assay Details
| Gene Symbol: Ube2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025356.4 | NP_079632.1 | 1 - 2 | 105 | 78 | |
| GenBank mRNA | AK004001.1 | - | 1 - 2 | 115 | 78 | |
| AK032885.1 | - | 1 - 2 | 115 | 78 | ||
| AK049906.1 | - | 1 - 2 | 106 | 78 | ||
| AK088448.1 | - | 1 - 2 | 75 | 78 | ||
| AK090099.1 | - | 117 | 78 | |||
| AK151594.1 | - | 1 - 2 | 126 | 78 | ||
| AK152231.1 | - | 1 - 2 | 126 | 78 | ||
| AK152541.1 | - | 1 - 2 | 99 | 78 | ||
| AK153111.1 | - | 1 - 2 | 127 | 78 | ||
| AK153410.1 | - | 1 - 2 | 126 | 78 | ||
| AK159546.1 | - | 1 - 2 | 101 | 78 | ||
| AK159963.1 | - | 1 - 2 | 99 | 78 | ||
| AK167077.1 | - | 1 - 2 | 115 | 78 | ||
| AK167650.1 | - | 1 - 2 | 103 | 78 | ||
| AK167801.1 | - | 1 - 2 | 127 | 78 | ||
| BC024351.1 | - | 1 - 2 | 110 | 78 | ||
| BC057941.1 | - | 1 - 2 | 86 | 5323636 | 78 | |
| BC098220.1 | - | 1 - 2 | 120 | 963757 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK004001.1,AK032885.1,AK049906.1,AK088448.1,AK151594.1,AK152231.1,AK152541.1,AK153111.1,AK153410.1,AK159546.1,AK159963.1,AK167077.1,AK167650.1,AK167801.1,BC024351.1,BC057941.1,BC098220.1,NM_025356.4 transcripts.
This assay is in the 5'UTR region of the AK004001.1,AK032885.1,AK049906.1,AK088448.1,AK151594.1,AK152231.1,AK152541.1,AK153111.1,AK153410.1,AK159546.1,AK159963.1,AK167077.1,AK167650.1,AK167801.1,BC024351.1,BC057941.1,BC098220.1,NM_025356.4 transcripts.
Set Membership: |
|

Ube2d3
Genomic Map