Assay Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025407.2 | NP_079683.2 | 3 - 4 | 366 | 99 | |
| GenBank mRNA | AK002292.1 | - | 3 - 4 | 307 | 99 | |
| AK004031.1 | - | 3 - 4 | 312 | 99 | ||
| AK010553.1 | - | 3 - 4 | 306 | 99 | ||
| AK013128.1 | - | 3 - 4 | 306 | 99 | ||
| AK151764.1 | - | 3 - 4 | 308 | 99 | ||
| AK160337.1 | - | 3 - 4 | 313 | 99 | ||
| AK167911.1 | - | 3 - 4 | 306 | 99 | ||
| AK168218.1 | - | 3 - 4 | 308 | 99 | ||
| AK177461.1 | - | 2 - 3 | 99 | |||
| AK184915.1 | - | 2 - 3 | 159 | 99 | ||
| BB649589.1 | - | 3 - 4 | 366 | 99 | ||
| BC030064.1 | - | 3 - 4 | 303 | 1400271 | 99 | |
| BY260300.1 | - | 3 - 4 | 339 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc1
Genomic Map