Assay Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025407.2 | NP_079683.2 | 4 - 5 | 495 | 86 | |
| GenBank mRNA | AK002292.1 | - | 4 - 5 | 436 | 86 | |
| AK004031.1 | - | 4 - 5 | 441 | 86 | ||
| AK010553.1 | - | 4 - 5 | 435 | 86 | ||
| AK013128.1 | - | 4 - 5 | 435 | 86 | ||
| AK151764.1 | - | 4 - 5 | 437 | 86 | ||
| AK160337.1 | - | 4 - 5 | 442 | 86 | ||
| AK167911.1 | - | 4 - 5 | 435 | 86 | ||
| AK168218.1 | - | 4 - 5 | 437 | 86 | ||
| AK177461.1 | - | 3 - 4 | 288 | 86 | ||
| AK184915.1 | - | 3 - 4 | 288 | 86 | ||
| AK185377.1 | - | 3 - 4 | 288 | 86 | ||
| BC030064.1 | - | 4 - 5 | 432 | 1400271 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc1
Genomic Map