Assay Details
| Gene Symbol: Uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025407.2 | NP_079683.2 | 8 - 9 | 1035 | 96 | |
| GenBank mRNA | AK004031.1 | - | 8 - 9 | 981 | 96 | |
| AK010553.1 | - | 8 - 9 | 975 | 96 | ||
| AK013128.1 | - | 8 - 9 | 96 | |||
| AK151764.1 | - | 8 - 9 | 977 | 96 | ||
| AK160337.1 | - | 8 - 9 | 982 | 96 | ||
| AK167911.1 | - | 8 - 9 | 975 | 96 | ||
| AK168218.1 | - | 8 - 9 | 977 | 96 | ||
| BC018405.1 | - | 1 - 2 | 88 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uqcrc1
Genomic Map