Assay Details
| Gene Symbol: Lgals2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025622.3 | NP_079898.2 | 1 - 2 | 63 | 73 | |
| GenBank mRNA | AK007364.1 | - | 1 - 2 | 60 | 73 | |
| AK008423.1 | - | 1 - 2 | 64 | 73 | ||
| AK008636.1 | - | 1 - 2 | 64 | 73 | ||
| AK008648.1 | - | 1 - 2 | 64 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007364.1, AK008423.1, AK008636.1, AK008648.1, NM_025622.3 transcripts.
Set Membership: |
|

Lgals2
Genomic Map