Assay Details
| Gene Symbol: Lgals2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025622.3 | NP_079898.2 | 3 - 4 | 302 | 82 | |
| GenBank mRNA | AK007364.1 | - | 3 - 4 | 299 | 82 | |
| AK008423.1 | - | 3 - 4 | 303 | 82 | ||
| AK008607.1 | - | 3 - 4 | 304 | 82 | ||
| AK008636.1 | - | 3 - 4 | 303 | 82 | ||
| AK008648.1 | - | 3 - 4 | 303 | 82 | ||
| BC027504.1 | - | 3 - 4 | 269 | 1434603 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665208 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30362771 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135791 | - | - |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AK008648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | AW456488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P563375 | NM_025622.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110065 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12038519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11907177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12038520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12038516 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12038517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30362799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11960075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12284748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11934089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11896132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12247936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12138570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12084056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12038517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12122549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12070239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12084065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12216551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12434385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11865033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11907178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11945108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11896133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12267773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12084027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30362764 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430426 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417078_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417079_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430425 | - | - |
| Mouse Genome 430 2.0 Array | 1417079_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417078_at | - | - |
| Mouse Genome 430 2.0 Array | 1417078_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417079_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107753_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107753_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014448.mm.1 | NM_025622.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014448.mm.1 | NM_025622.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014448.mm.1 | TC1500001849.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417078_PM_at | - | - |
| Mouse Expression Set 430 | 1417078_at | - | - |
| Mouse Expression Set 430 | 1417079_PM_s_at | - | - |
| Mouse Expression Set 430 | 1417079_s_at | - | - |
| Murine Genome U74v2 Set | 160221_at | - | - |
| Murine Genome U74v2 Set | 160221_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417079_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417078_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417078_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417079_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417079_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417078_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417079_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417078_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 604245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3610487 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641371 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2641371 | - | - |
| mouse-6 v1.1 (Illumina) | scl46981.3.21_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46981.3.21_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610487 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2641371 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641371 | - | - |

Lgals2
Genomic Map