Assay Details
Gene Symbol: Spryd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025716.2 | NP_079992.1 | 1 - 2 | 88 | 110 | |
GenBank mRNA | AK014623.1 | - | 1 - 2 | 93 | 110 | |
AK080726.1 | - | 1 - 2 | 102 | 110 | ||
AK082745.1 | - | 1 - 2 | 102 | 110 | ||
BC014757.1 | - | 1 - 2 | 88 | 4923277 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
