Assay Details
| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025859.3 | NP_080135.2 | 1 - 2 | 158 | 105 | |
| GenBank mRNA | AK009226.1 | - | 178 | 105 | ||
| AK010732.1 | - | 1 - 2 | 159 | 105 | ||
| AK018138.1 | - | 160 | 105 | |||
| AK088330.1 | - | 207 | 105 | |||
| AK167748.1 | - | 155 | 105 | |||
| BC096684.1 | - | 115 | 1248346 | 105 | ||
| BC116894.1 | - | 172 | 40126213 | 105 | ||
| BC116896.1 | - | 172 | 40126215 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009226.1, AK010732.1, AK018138.1, AK088330.1, AK167748.1, BC096684.1, BC116894.1, BC116896.1, NM_025859.3 transcripts.
Set Membership: |
|

Arl1
Genomic Map