Assay Details
| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025859.3 | NP_080135.2 | 3 - 4 | 372 | 102 | |
| GenBank mRNA | AK009226.1 | - | 392 | 102 | ||
| AK010732.1 | - | 3 - 4 | 373 | 102 | ||
| AK018138.1 | - | 374 | 102 | |||
| AK088330.1 | - | 421 | 102 | |||
| AK167748.1 | - | 369 | 102 | |||
| AK207710.1 | - | 2 - 3 | 146 | 102 | ||
| AK208964.1 | - | 2 - 3 | 151 | 102 | ||
| AK209287.1 | - | 2 - 3 | 102 | |||
| AK210028.1 | - | 2 - 3 | 182 | 102 | ||
| AK210540.1 | - | 2 - 3 | 178 | 102 | ||
| AK213037.1 | - | 2 - 3 | 134 | 102 | ||
| AK215653.1 | - | 2 - 3 | 160 | 102 | ||
| AK216046.1 | - | 2 - 3 | 151 | 102 | ||
| AK216240.1 | - | 2 - 3 | 154 | 102 | ||
| BC096684.1 | - | 329 | 1248346 | 102 | ||
| BC116894.1 | - | 386 | 40126213 | 102 | ||
| BC116896.1 | - | 386 | 40126215 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arl1
Genomic Map