Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026883.3 | NP_081159.1 | 3 - 4 | 257 | 112 | |
| GenBank mRNA | AK005211.1 | - | 3 - 4 | 649 | 112 | |
| AK150218.1 | - | 3 - 4 | 656 | 112 | ||
| BC019498.1 | - | 3 - 4 | 615 | 4460879 | 112 | |
| BC057658.1 | - | 3 - 4 | 296 | 3153717 | 112 | |
| BC061033.1 | - | 3 - 4 | 257 | 6590939 | 112 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam216a
Genomic Map