Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800013404.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013404.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013404.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013392.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013392.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013392.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013393.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013393.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013393.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013402.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013402.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013402.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013398.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013398.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013398.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013394.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013394.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013394.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013397.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013397.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013397.mm.1 |
TC0800001627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013399.mm.1 |
NM_026956.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013399.mm.1 |
NM_026956.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013399.mm.1 |
TC0800001627.mm.1 |
MTA TC |