Assay Details
| Gene Symbol: Snrpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027246.1 | NP_081522.1 | 2 - 3 | 240 | 88 | |
| GenBank mRNA | AK008240.1 | - | 2 - 3 | 241 | 88 | |
| AK135835.1 | - | 2 - 3 | 236 | 88 | ||
| BC066015.1 | - | 2 - 3 | 203 | 88 | ||
| BC100499.1 | - | 2 - 3 | 266 | 88 | ||
| BQ748807.1 | - | 2 - 3 | 212 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snrpf
Genomic Map