Assay Details
| Gene Symbol: Rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027332.3 | NP_081608.1 | 1 - 2 | 258 | 99 | |
| NM_027371.3 | NP_081647.2 | 1 - 2 | 258 | 99 | ||
| GenBank mRNA | AK008988.1 | - | 1 - 2 | 247 | 99 | |
| AK010079.1 | - | 1 - 2 | 255 | 99 | ||
| AK162073.1 | - | 1 - 2 | 261 | 99 | ||
| AK171035.1 | - | 1 - 2 | 239 | 99 | ||
| AK171079.1 | - | 1 - 2 | 247 | 99 | ||
| BC056165.1 | - | 1 - 2 | 209 | 5694049 | 99 | |
| BC089369.1 | - | 1 - 2 | 243 | 30550759 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC056165.1, BC089369.1 transcripts.
Set Membership: |
|

Rpf1
Genomic Map