Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300013723.mm.1 |
NM_027344.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013723.mm.1 |
NM_027344.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013723.mm.1 |
TC1300002139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013725.mm.1 |
NM_027344.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013725.mm.1 |
NM_027344.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013725.mm.1 |
TC1300002139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013727.mm.1 |
NM_027344.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013727.mm.1 |
NM_027344.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013727.mm.1 |
TC1300002139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013724.mm.1 |
NM_027344.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013724.mm.1 |
NM_027344.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013724.mm.1 |
TC1300002139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013722.mm.1 |
NM_027344.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013722.mm.1 |
NM_027344.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013722.mm.1 |
TC1300002139.mm.1 |
MTA TC |