Assay Details
| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027344.3 | NP_081620.2 | 5 - 6 | 710 | 76 | |
| GenBank mRNA | AK145382.1 | - | 5 - 6 | 714 | 76 | |
| BC147423.1 | - | 4 - 5 | 649 | 9056481 | 76 | |
| BC147424.1 | - | 4 - 5 | 649 | 9056499 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsll3
Genomic Map