Assay Details
| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027344.3 | NP_081620.2 | 6 - 7 | 875 | 99 | |
| GenBank mRNA | AK009935.1 | - | 1 - 2 | 270 | 99 | |
| AK145382.1 | - | 6 - 7 | 879 | 99 | ||
| BC147423.1 | - | 5 - 6 | 814 | 9056481 | 99 | |
| BC147424.1 | - | 5 - 6 | 814 | 9056499 | 99 | |
More Information
Set Membership: |
|

Ctsll3
Genomic Map