Assay Details
| Gene Symbol: Enkur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027728.2 | NP_082004.1 | 2 - 3 | 435 | 95 | |
| GenBank mRNA | AK017056.1 | - | 2 - 3 | 436 | 95 | |
| AY454124.1 | - | 2 - 3 | 433 | 95 | ||
| BC099519.1 | - | 2 - 3 | 334 | 30848009 | 95 | |
| BC132425.1 | - | 2 - 3 | 296 | 40130702 | 95 | |
| BC132451.1 | - | 2 - 3 | 296 | 40130728 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enkur
Genomic Map