Assay Details
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028623.5 | NP_082899.1 | 2 - 3 | 609 | 98 | |
GenBank mRNA | AK003744.1 | - | 2 - 3 | 421 | 98 | |
AK078116.1 | - | 2 - 3 | 610 | 98 | ||
BC027680.1 | - | 2 - 3 | 362 | 98 | ||
BC061036.1 | - | 2 - 3 | 395 | 6770352 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928180 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928179 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928178 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341978 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454545 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226844 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124786 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | AK078116.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | BC027680.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | BC061036.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | AK003744.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | AK078116.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | BC027680.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | BC061036.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417933 | AK078116.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417933 | NM_028623.5 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928202 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928215 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928191 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928182 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124786 | - | - |
Whole Mouse Genome G4122A | A_51_P417933 | AK078116.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P417933 | NM_028623.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | AK078116.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | BC027680.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | BC061036.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | NM_028623.5 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P59206 | AK003744.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122385 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124786 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14276978 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14276971 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14181766 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14276977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928180 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928178 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31928179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14185025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14441702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14276987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14350669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14322003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14350658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14418848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14167820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14452819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14452824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14185024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14185026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14350661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14350668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14167818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928145 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14322002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31928182 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10465002 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465004 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465001 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465003 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427910_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10464999 | - | - |
Mouse Genome 430 2.0 Array | 1427910_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427910_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73720_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73720_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900008509.mm.1 | NM_028623.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008509.mm.1 | NM_028623.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008509.mm.1 | TC1900000959.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900008510.mm.1 | NM_028623.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008510.mm.1 | NM_028623.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008510.mm.1 | TC1900000959.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427910_PM_at | - | - |
Mouse Expression Set 430 | 1427910_at | - | - |
Mouse Genome 430A 2.0 Array | 1427910_at | - | - |
Mouse Genome 430A 2.0 Array | 1427910_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427910_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427910_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 468180 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4760465 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004760465 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247676 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247676 | - | - |
mouse-6 v1.1 (Illumina) | scl53490.4_84-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4760465 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl53490.4_84-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760465 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1247676 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2872851 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2872851 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247676 | - | - |
