Assay Details
| Gene Symbol: Pcdhga8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033591.3 | NP_291069.1 | 1 - 2 | 2618 | 67 | |
| GenBank mRNA | AK220461.1 | - | 1 - 2 | 2571 | 67 | |
| AY013802.1 | - | 1 - 2 | 2431 | 67 | ||
| BC054555.1 | - | 1 - 2 | 2579 | 5698395 | 67 | |
| BC054787.1 | - | 1 - 2 | 2618 | 6825772 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcdhga8
Genomic Map