Assay Details
Gene Symbol: Pcdhga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033594.2 | NP_291072.1 | 1 - 2 | 2596 | 82 | |
GenBank mRNA | AY013794.1 | - | 1 - 2 | 2428 | 82 | |
BC059837.1 | - | 1 - 2 | 2527 | 5719590 | 82 | |
BC064675.1 | - | 1 - 2 | 2596 | 6406939 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
