Assay Details
| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053164.3 | NP_444394.1 | 2 - 3 | 285 | 83 | |
| GenBank mRNA | AB049657.1 | - | 2 - 3 | 246 | 83 | |
| AK015357.1 | - | 2 - 3 | 266 | 83 | ||
| AK076204.1 | - | 2 - 3 | 244 | 83 | ||
| BC027676.1 | - | 2 - 3 | 225 | 83 | ||
| BC048733.1 | - | 2 - 3 | 276 | 6512868 | 83 | |
| BC065812.1 | - | 2 - 3 | 232 | 83 | ||
| BY251956.1 | - | 2 - 3 | 286 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135816 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135813 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135819 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135815 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135814 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174895 | AK015357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174895 | AK076204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174895 | BC027676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174895 | BC048733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174895 | BC065812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605931 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115459 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174895 | AK015357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174895 | AK076204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174895 | BC027676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174895 | BC048733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174895 | BC065812.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P374351 | AK015357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P374351 | AK076204.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P374351 | BC027676.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P374351 | BC048733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P374351 | BC065812.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32135818 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115459 | - | - |
| Whole Mouse Genome G4122A | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | AK015357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | AK076204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC027676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC048733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P255441 | BC065812.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374351 | AK015357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374351 | AK076204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374351 | BC027676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374351 | BC048733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374351 | BC065812.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101827 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115459 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14265539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14265520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14265536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14265526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135814 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135815 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135813 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135816 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135819 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32135817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32135751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14265531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14265552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14265520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32135809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32135818 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468029 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415681_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468022 | - | - |
| Mouse Genome 430 2.0 Array | 1415681_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415681_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94067_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94067_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900013918.mm.1 | NM_053164.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013918.mm.1 | NM_053164.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013918.mm.1 | TC1900001569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900013917.mm.1 | NM_053164.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013917.mm.1 | NM_053164.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013917.mm.1 | TC1900001569.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415681_PM_at | - | - |
| Mouse Expression Set 430 | 1415681_at | - | - |
| Mouse Expression Set 430 | 1415681_PM_at | - | - |
| Mouse Expression Set 430 | 1415681_at | - | - |
| Murine Genome U74v2 Set | 99674_PM_at | - | - |
| Murine Genome U74v2 Set | 99674_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415681_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415681_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415681_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415681_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415681_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415681_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 360014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 360014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5360673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598222 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2633805 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2598222 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2633805 | - | - |
| mouse-6 v1.1 (Illumina) | scl52435.3.1_22-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000468.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360014 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000468.1_25-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52435.3.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360014 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2633805 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2598222 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2598222 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2633805 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52435.3.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000468.1_25-S | - | - |

Mrpl43
Genomic Map