Assay Details
Gene Symbol: Btnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079835.2 | NP_524574.1 | 3 - 4 | 678 | 100 | |
GenBank mRNA | AK136521.1 | - | 3 - 4 | 680 | 100 | |
BC141103.1 | - | 3 - 4 | 568 | 9055715 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
