Assay Details
| Gene Symbol: Rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144948.5 | NP_659197.2 | 1 - 2 | 172 | 102 | |
| GenBank mRNA | AF458961.1 | - | 1 - 2 | 136 | 102 | |
| AK005200.1 | - | 1 - 2 | 148 | 102 | ||
| AK006755.1 | - | 1 - 2 | 137 | 102 | ||
| AK008645.1 | - | 1 - 2 | 132 | 102 | ||
| AK011223.1 | - | 1 - 2 | 154 | 102 | ||
| AK015662.1 | - | 1 - 2 | 173 | 102 | ||
| AK145381.1 | - | 1 - 2 | 136 | 102 | ||
| AK151056.1 | - | 1 - 2 | 151 | 102 | ||
| AK151827.1 | - | 1 - 2 | 167 | 102 | ||
| BC054774.1 | - | 1 - 2 | 139 | 6807637 | 102 | |
| BC116931.1 | - | 1 - 2 | 121 | 40126250 | 102 | |
| BC119038.1 | - | 1 - 2 | 121 | 8733787 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm7
Genomic Map