Assay Details
| Gene Symbol: Rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144948.5 | NP_659197.2 | 2 - 3 | 336 | 93 | |
| NR_037589.1 | 3 - 4 | 441 | 93 | |||
| GenBank mRNA | AF458961.1 | - | 2 - 3 | 300 | 93 | |
| AK005200.1 | - | 2 - 3 | 312 | 93 | ||
| AK006755.1 | - | 2 - 3 | 301 | 93 | ||
| AK008645.1 | - | 2 - 3 | 296 | 93 | ||
| AK011223.1 | - | 2 - 3 | 318 | 93 | ||
| AK145381.1 | - | 2 - 3 | 300 | 93 | ||
| AK151056.1 | - | 2 - 3 | 315 | 93 | ||
| AK151827.1 | - | 2 - 3 | 331 | 93 | ||
| AK153504.1 | - | 3 - 4 | 405 | 93 | ||
| AK182663.1 | - | 1 - 2 | 93 | |||
| AK213555.1 | - | 1 - 2 | 153 | 93 | ||
| AK216808.1 | - | 1 - 2 | 153 | 93 | ||
| BC054774.1 | - | 2 - 3 | 303 | 6807637 | 93 | |
| BC116931.1 | - | 2 - 3 | 285 | 40126250 | 93 | |
| BC119038.1 | - | 2 - 3 | 285 | 8733787 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm7
Genomic Map