Assay Details
Gene Symbol: Spsb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145134.3 | NP_660116.1 | 2 - 3 | 1011 | 114 | |
GenBank mRNA | AF403039.2 | - | 2 - 3 | 1011 | 114 | |
AK142195.1 | - | 2 - 3 | 891 | 114 | ||
BC023083.1 | - | 1 - 2 | 745 | 4953500 | 114 | |
BC055705.1 | - | 2 - 3 | 879 | 6412566 | 114 | |
BC094332.1 | - | 2 - 3 | 1474 | 6817091 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
