Assay Details
| Gene Symbol: Igflr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145580.2 | NP_663555.2 | 1 - 2 | 40 | 79 | |
| GenBank mRNA | AK154129.1 | - | 1 - 2 | 41 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154129.1, NM_145580.2 transcripts.
Set Membership: |
|

Igflr1
Genomic Map