Assay Details
Gene Symbol: Igflr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145580.2 | NP_663555.2 | 2 - 3 | 258 | 87 | |
GenBank mRNA | AK154129.1 | - | 2 - 3 | 259 | 87 | |
BC019733.1 | - | 1 - 2 | 3676256 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
