Assay Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146184.4 | NP_666296.1 | 1 | 354 | 91 | |
| GenBank mRNA | AK042388.1 | - | 1 | 337 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042388.1, NM_146184.4 transcripts.
Set Membership: |
|

B3gnt8
Genomic Map