Assay Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013921.3 | NP_038949.2 | 5 - 6 | 730 | 99 | |
| GenBank mRNA | AJ243866.1 | - | 5 - 6 | 693 | 99 | |
| AK008263.1 | - | 4 - 5 | 99 | |||
| BC010970.1 | - | 5 - 6 | 723 | 4216644 | 99 | |
| BC040348.1 | - | 5 - 6 | 730 | 4206523 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294031 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173784 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173784 | - | - |
| Whole Mouse Genome G4122A | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241935 | AJ243866.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123031 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097759 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13097751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13548032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13097741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13083717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13535243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13375197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13548029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13571173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13381073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13078958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13523724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13083715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13305505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13195428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13548031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13460289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13097751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13097755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13097740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10448407 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448408 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448405 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449952_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448402 | - | - |
| Mouse Genome 430 2.0 Array | 1449952_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449952_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30943_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30943_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014120.mm.1 | NM_013921.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014120.mm.1 | NM_013921.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014120.mm.1 | TC1700001611.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014121.mm.1 | NM_013921.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014121.mm.1 | NM_013921.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014121.mm.1 | TC1700001611.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014122.mm.1 | NM_013921.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014122.mm.1 | NM_013921.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014122.mm.1 | TC1700001611.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449952_s_at | - | - |
| Mouse Expression Set 430 | 1449952_PM_s_at | - | - |
| Murine Genome U74v2 Set | 93709_PM_at | - | - |
| Murine Genome U74v2 Set | 93709_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449952_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449952_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449952_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449952_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 463972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6620575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258386 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258386 | - | - |
| mouse-6 v1.1 (Illumina) | scl50223.6.1_69-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50223.6.1_69-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1258386 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258386 | - | - |

Prss30
Genomic Map