Assay Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016668.3 | NP_057877.1 | 1 - 2 | 290 | 119 | |
| GenBank mRNA | AK077108.1 | - | 1 - 2 | 97 | 119 | |
| AK145668.1 | - | 1 - 2 | 119 | |||
| AK149431.1 | - | 1 - 2 | 99 | 119 | ||
| AK149457.1 | - | 1 - 2 | 97 | 119 | ||
| AK166754.1 | - | 1 - 2 | 99 | 119 | ||
| AK166818.1 | - | 1 - 2 | 98 | 119 | ||
| BC037004.1 | - | 1 - 2 | 92 | 5100429 | 119 | |
| BC093510.1 | - | 1 - 2 | 65 | 5101184 | 119 | |
| BC110307.1 | - | 1 - 2 | 493 | 5310035 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145668.1,AK149431.1,AK149457.1,AK166754.1,AK166818.1,BC037004.1,BC093510.1,BC110307.1,NM_016668.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bhmt
Genomic Map