Assay Details
Gene Symbol: Olfr1076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146406.2 | NP_666518.2 | 1 | 852 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P680637 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P680637 | - | - |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P680637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402601 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21402600 | - | - |
Whole Mouse Genome G4122A | A_52_P680637 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109882 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112027 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01719048 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02193995 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02193990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21402663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01650550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402650 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01650529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01246673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01650531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21402637 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473506 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473505 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 7100324 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7100324 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007100324 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258776 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258776 | - | - |
mouse-6 v1.1 (Illumina) | scl20674.1.22_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7100324 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20674.1.22_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100324 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1258776 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258776 | - | - |
