Assay Details
| Gene Symbol: Olfr215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146446.1 | NP_666657.1 | 1 | 448 | 90 | |
| GenBank mRNA | BC125561.1 | - | 1 | 535 | 40130076 | 90 |
| BC125563.1 | - | 1 | 535 | 40130078 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017227 | NM_146446.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017227 | BC125561.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017227 | BC125563.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18760 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124878 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017227 | NM_146446.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017227 | BC125561.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017227 | BC125563.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P283615 | NM_146446.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P283615 | BC125561.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P283615 | BC125563.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24777219 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124878 | - | - |
| Whole Mouse Genome G4122A | A_52_P283615 | NM_146446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P283615 | BC125561.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P283615 | BC125563.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133286 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103042 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05339005 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05622599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05338998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05338991 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05622604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05339000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24777215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05338995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05337466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05316894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05338999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05622609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24777198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547172 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547171 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600022917.mm.1 | TC0600002976.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3800041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2673515 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2673515 | - | - |
| mouse-6 v1.1 (Illumina) | scl28511.1.1_86-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28511.1.1_86-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2673515 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2673515 | - | - |

Olfr215
Genomic Map