Assay Details
Gene Symbol: Olfr360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146622.1 | NP_666833.1 | 1 | 651 | 107 | |
GenBank mRNA | BC110546.2 | - | 1 | 649 | 107 | |
BC125238.1 | - | 1 | 649 | 107 | ||
BC125239.1 | - | 1 | 650 | 107 | ||
BC126918.1 | - | 1 | 650 | 107 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040285 | BC126918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040285 | NM_146622.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040285 | BC110546.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040285 | BC125238.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040285 | BC125239.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149793 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040285 | BC126918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040285 | NM_146622.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040285 | BC110546.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040285 | BC125238.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040285 | BC125239.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165575 | BC126918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165575 | NM_146622.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165575 | BC110546.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165575 | BC125238.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165575 | BC125239.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140825 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140801 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140828 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140789 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149793 | - | - |
Whole Mouse Genome G4122A | A_52_P165575 | BC126918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165575 | NM_146622.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165575 | BC110546.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165575 | BC125238.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165575 | BC125239.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123233 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149793 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01778983 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01200750 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01200749 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01778981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01200750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02074147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140827 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02220877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01834590 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01476520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01200743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01200749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01312274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01567930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140788 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10482132 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471773 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10482131 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471772 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 735694 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6650091 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650091 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224535 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224535 | - | - |
mouse-6 v1.1 (Illumina) | scl21006.1.1_59-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650091 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21006.1.1_59-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650091 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1224535 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3006758 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224535 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3006758 | - | - |
