| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200012287.mm.1 |
NM_146884.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012287.mm.1 |
NM_146884.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012287.mm.1 |
TC0200001652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012290.mm.1 |
NM_146884.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012290.mm.1 |
NM_146884.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012290.mm.1 |
TC0200001652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012291.mm.1 |
NM_146884.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012291.mm.1 |
NM_146884.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012291.mm.1 |
TC0200001652.mm.1 |
MTA TC |