| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200034022.mm.1 |
NM_146891.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034022.mm.1 |
NM_146891.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034022.mm.1 |
TC0200004046.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034001.mm.1 |
NM_146899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034001.mm.1 |
NM_146899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034001.mm.1 |
TC0200004039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034021.mm.1 |
NM_146891.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034021.mm.1 |
NM_146891.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034021.mm.1 |
TC0200004046.mm.1 |
MTA TC |