Assay Details
| Gene Symbol: U2af1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170760.3 | NP_739566.1 | 1 - 2 | 57 | 100 | |
| GenBank mRNA | BC060972.1 | - | 1 - 2 | 57 | 6593404 | 100 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC060972.1,NM_170760.3 transcripts.
Set Membership: |
|

U2af1l4
Genomic Map