Assay Details
| Gene Symbol: U2af1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170760.3 | NP_739566.1 | 7 - 8 | 589 | 61 | |
| GenBank mRNA | AF419339.1 | - | 7 - 8 | 573 | 61 | |
| BC060972.1 | - | 7 - 8 | 589 | 6593404 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25032192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25032193 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138090 | BC060972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138090 | NM_170760.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138090 | BC051477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211538 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136562 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138090 | BC060972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138090 | NM_170760.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138090 | BC051477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032204 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136562 | - | - |
| Whole Mouse Genome G4122A | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509140 | BC060972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509140 | NM_170760.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509140 | AF419339.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136562 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06035647 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06035662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06035665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25032192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25032193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06035655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032195 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551974 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551980 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551981 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551971 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233073_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233073_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700005062.mm.1 | TC0700000567.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005062.mm.1 | NM_170760.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005062.mm.1 | NM_170760.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005055.mm.1 | TC0700000567.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005055.mm.1 | NM_170760.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005055.mm.1 | NM_170760.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005061.mm.1 | TC0700000567.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005061.mm.1 | NM_170760.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005061.mm.1 | NM_170760.3 | Refseq |
| Mouse Expression Set 430 | 1434305_PM_at | - | - |
| Mouse Expression Set 430 | 1434305_at | - | - |
| Murine Genome U74v2 Set | 160432_PM_at | - | - |
| Murine Genome U74v2 Set | 160432_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434305_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434305_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 601904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160302 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160302 | - | - |
| mouse-6 v1.1 (Illumina) | GI_40254199-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160302 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160302 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_40254199-S | - | - |

U2af1l4
Genomic Map