Assay Details
Gene Symbol: Dpp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172624.3 | NP_766212.2 | 11 - 12 | 1498 | 90 | |
GenBank mRNA | AK039652.1 | - | 10 - 11 | 1591 | 90 | |
AK050021.1 | - | 11 - 12 | 1440 | 90 | ||
AK050023.1 | - | 11 - 12 | 1500 | 90 | ||
AK078301.1 | - | 11 - 12 | 1446 | 90 | ||
AK131178.1 | - | 8 - 9 | 3361 | 90 | ||
BC057631.1 | - | 11 - 12 | 1497 | 4191886 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
