Assay Details
| Gene Symbol: Prpf38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172697.3 | NP_766285.2 | 2 - 3 | 515 | 104 | |
| GenBank mRNA | AK075925.1 | - | 2 - 3 | 507 | 104 | |
| AK169741.1 | - | 2 - 3 | 468 | 104 | ||
| BC024734.1 | - | 2 - 3 | 488 | 104 | ||
| BC025853.1 | - | 2 - 3 | 417 | 104 | ||
| BC139277.1 | - | 2 - 3 | 453 | 8862299 | 104 | |
| BC139305.1 | - | 2 - 3 | 451 | 8862327 | 104 | |
| CT010186.1 | - | 2 - 3 | 293 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf38a
Genomic Map