Assay Details
| Gene Symbol: Prpf38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172697.3 | NP_766285.2 | 5 - 6 | 824 | 103 | |
| GenBank mRNA | AK075925.1 | - | 5 - 6 | 816 | 103 | |
| AK169741.1 | - | 5 - 6 | 777 | 103 | ||
| BC024734.1 | - | 5 - 6 | 797 | 103 | ||
| BC025853.1 | - | 5 - 6 | 726 | 103 | ||
| BC139277.1 | - | 5 - 6 | 762 | 8862299 | 103 | |
| BC139305.1 | - | 5 - 6 | 760 | 8862327 | 103 | |
| CT010186.1 | - | 5 - 6 | 602 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf38a
Genomic Map