Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172715.3 | NP_766303.1 | 3 - 4 | 493 | 112 | |
| GenBank mRNA | AK030171.1 | - | 4 - 5 | 678 | 112 | |
| AK138410.1 | - | 2 - 3 | 653 | 112 | ||
| AK160261.1 | - | 3 - 4 | 438 | 112 | ||
| BC096769.1 | - | 3 - 4 | 426 | 30101305 | 112 | |
| BC138228.1 | - | 2 - 3 | 246 | 8861247 | 112 | |
| BC145669.1 | - | 2 - 3 | 246 | 40130972 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat9
Genomic Map