Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172715.3 | NP_766303.1 | 7 - 8 | 1028 | 98 | |
| GenBank mRNA | AK030171.1 | - | 8 - 9 | 1213 | 98 | |
| AK138410.1 | - | 6 - 7 | 1188 | 98 | ||
| AK160261.1 | - | 7 - 8 | 973 | 98 | ||
| BC096769.1 | - | 7 - 8 | 961 | 30101305 | 98 | |
| BC138228.1 | - | 6 - 7 | 781 | 8861247 | 98 | |
| BC145669.1 | - | 6 - 7 | 781 | 40130972 | 98 | |
| CN719213.1 | - | 3 - 4 | 239 | 98 | ||
More Information
Set Membership: |
|

Agpat9
Genomic Map