Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172715.3 | NP_766303.1 | 9 - 10 | 1197 | 102 | |
| GenBank mRNA | AK030171.1 | - | 10 - 11 | 1382 | 102 | |
| AK138410.1 | - | 8 - 9 | 1357 | 102 | ||
| AK160261.1 | - | 9 - 10 | 1142 | 102 | ||
| BC096769.1 | - | 9 - 10 | 1130 | 30101305 | 102 | |
| BC138228.1 | - | 8 - 9 | 950 | 8861247 | 102 | |
| BC145669.1 | - | 8 - 9 | 950 | 40130972 | 102 | |
| CN719213.1 | - | 5 - 6 | 408 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat9
Genomic Map