Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200043428.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043429.mm.1 |
NM_173397.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043429.mm.1 |
NM_173397.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043429.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043432.mm.1 |
NM_173397.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043432.mm.1 |
NM_173397.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043432.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043430.mm.1 |
NM_173397.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043430.mm.1 |
NM_173397.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043430.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043431.mm.1 |
NM_173397.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043431.mm.1 |
NM_173397.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043431.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043426.mm.1 |
TC0200005091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043427.mm.1 |
TC0200005091.mm.1 |
MTA TC |