Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00000339.mm.1 |
NM_173747.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000339.mm.1 |
NM_173747.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000339.mm.1 |
TC0X00000063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000334.mm.1 |
NM_173747.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000334.mm.1 |
NM_173747.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000334.mm.1 |
TC0X00000063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000336.mm.1 |
NM_173747.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000336.mm.1 |
NM_173747.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000336.mm.1 |
TC0X00000063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000335.mm.1 |
NM_173747.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000335.mm.1 |
NM_173747.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000335.mm.1 |
TC0X00000063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000338.mm.1 |
NM_173747.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000338.mm.1 |
NM_173747.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000338.mm.1 |
TC0X00000063.mm.1 |
MTA TC |