Assay Details
| Gene Symbol: Hist1h3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178207.2 | NP_835514.1 | 1 | 48 | 64 | |
| GenBank mRNA | AK155722.1 | - | 49 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155722.1, NM_178207.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874606 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874577 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121521 | NM_178207.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121521 | AK155722.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94009 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121521 | NM_178207.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121521 | AK155722.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10517870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10517868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10910119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10760754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10910135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10517866 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10760755 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10517872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10662000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10517862 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494403 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403941 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408202 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408083 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408239 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404028 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404065 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408246 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404049 | - | - |
| Mouse Genome 430 2.0 Array | 1443475_at | - | - |
| Mouse Genome 430 2.0 Array | 1443475_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300009852.mm.1 | NM_178207.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009852.mm.1 | NM_178207.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009852.mm.1 | TC1300001561.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009850.mm.1 | NM_178207.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009850.mm.1 | NM_178207.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009850.mm.1 | TC1300001561.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009851.mm.1 | NM_178207.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009851.mm.1 | NM_178207.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009851.mm.1 | TC1300001561.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460314_s_at | - | - |
| Mouse Expression Set 430 | 1427864_at | - | - |
| Mouse Expression Set 430 | 1460314_PM_s_at | - | - |
| Mouse Expression Set 430 | 1427864_PM_at | - | - |
| Mouse Expression Set 430 | 1443475_PM_at | - | - |
| Mouse Expression Set 430 | 1443475_at | - | - |
| Murine Genome U74v2 Set | 94756_PM_at | - | - |
| Murine Genome U74v2 Set | 96416_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93023_f_at | - | - |
| Murine Genome U74v2 Set | 94756_at | - | - |
| Murine Genome U74v2 Set | 96416_f_at | - | - |
| Murine Genome U74v2 Set | 93023_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7100270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7100270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007100270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697777 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698519 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697777 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698519 | - | - |
| mouse-6 v1.1 (Illumina) | scl0319153.1_241-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7100270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319153.1_241-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100270 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698519 | - | - |

Hist1h3i
Genomic Map