Assay Details
Gene Symbol: Mtmr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023858.3 | NP_076347.3 | 1 | 49 | 80 | |
NR_024025.1 | 1 | 49 | 80 | |||
GenBank mRNA | AK020065.1 | - | 1 | 50 | 80 | |
AY055832.1 | - | 1 | 820 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK020065.1, AY055832.1, NM_023858.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
