Assay Details
| Gene Symbol: Cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025415.3 | NP_079691.1 | 1 | 49 | 63 | |
| GenBank mRNA | DV661312.1 | - | 1 | 49 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_025415.3 transcripts.
Set Membership: |
|

Cks2
Genomic Map